The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001120.4 | NP_001111.3 | 8 - 9 | 1340 | 92 |
NM_001146069.1 | NP_001139541.1 | 9 - 10 | 1138 | 92 | |
XM_005247927.3 | XP_005247984.1 | 9 - 10 | 1133 | 92 | |
GenBank mRNA | BC001502.2 | - | 9 - 10 | 1064 | 92 |
BC014979.1 | - | 9 - 10 | 1067 | 92 | |
L11669.1 | - | 8 - 9 | 1146 | 92 |