Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COG7
Entrez Gene ID
91949
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
CDG2E
Chromosome Location
Chr.16: 23388493 - 23453215 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153603.3 NP_705831.1 3 - 4 635 69
XM_017023870.1 XP_016879359.1 3 - 4 678 69
GenBank mRNA BC000549.2 - 3 - 4 590 69
BC037563.1 - 3 - 4 624 69
KU178837.1 - 3 - 4 438 69

View Full Product Details