Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMOD4
Entrez Gene ID
29765
Gene Name
tropomodulin 4
Gene Aliases
SK-TMOD
Chromosome Location
Chr.1: 151169986 - 151176071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013353.2 NP_037485.2 6 - 7 810 68
XM_011509449.1 XP_011507751.1 6 - 7 692 68
XM_017001089.1 XP_016856578.1 5 - 6 1427 68
GenBank mRNA AF165217.1 - 6 - 7 687 68
AF177173.2 - 6 - 7 755 68
AK092700.1 - 6 - 7 676 68
AK300675.1 - 4 - 5 513 68
AK313280.1 - 6 - 7 719 68
BC017810.1 - 6 - 7 720 68

View Full Product Details