The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024586.5 | NP_078862.4 | 21 - 22 | 2141 | 115 |
NM_148904.3 | NP_683702.1 | 17 - 18 | 1858 | 115 | |
NM_148905.3 | NP_683703.1 | 16 - 17 | 1762 | 115 | |
NM_148906.2 | NP_683704.2 | 20 - 21 | 2090 | 115 | |
NM_148907.2 | NP_683705.1 | 17 - 18 | 1735 | 115 | |
NM_148908.3 | NP_683706.3 | 20 - 21 | 2102 | 115 | |
NM_148909.3 | NP_683707.3 | 21 - 22 | 2171 | 115 | |
NR_036662.1 | 20 - 21 | 2062 | 115 | ||
XM_006710318.2 | XP_006710381.1 | 20 - 21 | 1901 | 115 | |
XM_006710320.3 | XP_006710383.1 | 18 - 19 | 1738 | 115 | |
XM_006710321.2 | XP_006710384.1 | 17 - 18 | 1734 | 115 | |
XM_006710322.3 | XP_006710385.1 | 18 - 19 | 1895 | 115 | |
XM_006710323.2 | XP_006710386.1 | 17 - 18 | 1803 | 115 | |
XM_006710324.3 | XP_006710387.1 | 19 - 20 | 1813 | 115 | |
XM_006710325.3 | XP_006710388.1 | 18 - 19 | 1783 | 115 | |
XM_006710326.2 | XP_006710389.1 | 16 - 17 | 1785 | 115 | |
XM_011540599.2 | XP_011538901.1 | 22 - 23 | 1995 | 115 | |
XM_011540600.2 | XP_011538902.1 | 21 - 22 | 1962 | 115 | |
XM_011540601.2 | XP_011538903.1 | 20 - 21 | 1923 | 115 | |
XM_011540602.2 | XP_011538904.1 | 21 - 22 | 1897 | 115 | |
XM_011540603.2 | XP_011538905.1 | 21 - 22 | 1921 | 115 | |
XM_011540604.2 | XP_011538906.1 | 18 - 19 | 1696 | 115 | |
XM_017000218.1 | XP_016855707.1 | 19 - 20 | 1862 | 115 | |
XM_017000219.1 | XP_016855708.1 | 20 - 21 | 1882 | 115 | |
XM_017000220.1 | XP_016855709.1 | 17 - 18 | 1623 | 115 | |
XM_017000221.1 | XP_016855710.1 | 17 - 18 | 1574 | 115 | |
XM_017000222.1 | XP_016855711.1 | 21 - 22 | 2107 | 115 | |
XM_017000223.1 | XP_016855712.1 | 21 - 22 | 2105 | 115 | |
XM_017000224.1 | XP_016855713.1 | 18 - 19 | 2050 | 115 | |
XM_017000225.1 | XP_016855714.1 | 19 - 20 | 1901 | 115 | |
GenBank mRNA | AF392445.1 | - | 21 - 22 | 1915 | 115 |
AK022554.1 | - | 16 - 17 | 1542 | 115 | |
AK027535.1 | - | 20 - 21 | 1872 | 115 | |
AK027707.1 | - | 17 - 18 | 1698 | 115 | |
AK056617.1 | - | 21 - 22 | 1960 | 115 | |
AK074392.1 | - | 7 - 8 | 674 | 115 | |
AK091703.1 | - | 20 - 21 | 1844 | 115 | |
AK124037.1 | - | 12 - 13 | 2388 | 115 | |
AK128043.1 | - | 23 - 24 | 2365 | 115 | |
AK297656.1 | - | 18 - 19 | 1888 | 115 | |
AK315167.1 | - | 17 - 18 | 1731 | 115 | |
AY178997.1 | - | 20 - 21 | 1864 | 115 | |
BC006160.2 | - | 1 - 2 | 74 | 115 | |
BC025978.2 | - | 17 - 18 | 1768 | 115 | |
CR457288.1 | - | 14 - 15 | 1409 | 115 |