Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OSBPL9
Entrez Gene ID
114883
Gene Name
oxysterol binding protein like 9
Gene Aliases
ORP-9, ORP9
Chromosome Location
Chr.1: 51616874 - 51789219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024586.5 NP_078862.4 13 - 14 1286 96
NM_148904.3 NP_683702.1 9 - 10 1003 96
NM_148905.3 NP_683703.1 8 - 9 907 96
NM_148906.2 NP_683704.2 12 - 13 1235 96
NM_148907.2 NP_683705.1 9 - 10 880 96
NM_148908.3 NP_683706.3 12 - 13 1247 96
NM_148909.3 NP_683707.3 13 - 14 1316 96
NR_036662.1 12 - 13 1207 96
XM_006710318.2 XP_006710381.1 12 - 13 1046 96
XM_006710320.3 XP_006710383.1 10 - 11 883 96
XM_006710321.2 XP_006710384.1 9 - 10 879 96
XM_006710322.3 XP_006710385.1 10 - 11 1040 96
XM_006710323.2 XP_006710386.1 9 - 10 948 96
XM_006710324.3 XP_006710387.1 11 - 12 958 96
XM_006710325.3 XP_006710388.1 10 - 11 928 96
XM_006710326.2 XP_006710389.1 8 - 9 930 96
XM_011540599.2 XP_011538901.1 14 - 15 1140 96
XM_011540600.2 XP_011538902.1 13 - 14 1107 96
XM_011540601.2 XP_011538903.1 12 - 13 1068 96
XM_011540602.2 XP_011538904.1 13 - 14 1042 96
XM_011540603.2 XP_011538905.1 13 - 14 1066 96
XM_011540604.2 XP_011538906.1 10 - 11 841 96
XM_017000218.1 XP_016855707.1 11 - 12 1007 96
XM_017000219.1 XP_016855708.1 12 - 13 1027 96
XM_017000220.1 XP_016855709.1 9 - 10 768 96
XM_017000221.1 XP_016855710.1 9 - 10 719 96
XM_017000222.1 XP_016855711.1 13 - 14 1252 96
XM_017000223.1 XP_016855712.1 13 - 14 1250 96
XM_017000224.1 XP_016855713.1 10 - 11 1195 96
XM_017000225.1 XP_016855714.1 11 - 12 1046 96
GenBank mRNA AB208851.1 - 2 - 3 553 96
AF392445.1 - 13 - 14 1060 96
AK022554.1 - 8 - 9 687 96
AK027535.1 - 12 - 13 1017 96
AK027707.1 - 9 - 10 843 96
AK056617.1 - 13 - 14 1105 96
AK091703.1 - 12 - 13 989 96
AK124037.1 - 4 - 5 1533 96
AK128043.1 - 15 - 16 1510 96
AK297656.1 - 10 - 11 1033 96
AK315167.1 - 9 - 10 879 96
AY178997.1 - 12 - 13 1009 96
BC025978.2 - 9 - 10 913 96
CR457288.1 - 6 - 7 554 96

View Full Product Details