The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176877.2 | NP_795352.2 | 17 - 18 | 2227 | 116 |
XM_005270341.3 | XP_005270398.1 | 17 - 18 | 2225 | 116 | |
XM_005270347.1 | XP_005270404.1 | 17 - 18 | 2225 | 116 | |
XM_006710278.3 | XP_006710341.1 | 17 - 18 | 2225 | 116 | |
XM_011540462.2 | XP_011538764.1 | 17 - 18 | 2225 | 116 | |
XM_011540463.2 | XP_011538765.1 | 17 - 18 | 2413 | 116 | |
XM_011540464.2 | XP_011538766.1 | 17 - 18 | 2225 | 116 | |
XM_011540465.2 | XP_011538767.1 | 17 - 18 | 2225 | 116 | |
XM_011540466.2 | XP_011538768.1 | 17 - 18 | 2225 | 116 | |
XM_011540467.2 | XP_011538769.1 | 17 - 18 | 2225 | 116 | |
XM_011540468.2 | XP_011538770.1 | 17 - 18 | 2225 | 116 | |
XM_011540469.2 | XP_011538771.1 | 17 - 18 | 2225 | 116 | |
XM_011540470.1 | XP_011538772.1 | 17 - 18 | 2225 | 116 | |
XM_016999998.1 | XP_016855487.1 | 17 - 18 | 2225 | 116 | |
XM_016999999.1 | XP_016855488.1 | 17 - 18 | 2225 | 116 | |
XM_017000000.1 | XP_016855489.1 | 12 - 13 | 1471 | 116 | |
GenBank mRNA | AB044807.1 | - | 4 - 5 | 506 | 116 |
AF397170.1 | - | 16 - 17 | 2113 | 116 | |
AJ001306.1 | - | 17 - 18 | 2227 | 116 | |
AJ224747.1 | - | 17 - 18 | 2227 | 116 | |
AJ224748.1 | - | 17 - 18 | 2227 | 116 | |
AK304355.1 | - | 12 - 13 | 1584 | 116 | |
AK307713.1 | - | 17 - 18 | 2224 | 116 | |
BC142661.1 | - | 17 - 18 | 2210 | 116 | |
BC142723.1 | - | 17 - 18 | 2210 | 116 | |
BC144135.1 | - | 17 - 18 | 2179 | 116 |