Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8A
Entrez Gene ID
731
Gene Name
complement component 8 alpha subunit
Gene Aliases
-
Chromosome Location
Chr.1: 56854770 - 56918221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000562.2 NP_000553.1 1 - 2 201 107
XM_011542079.2 XP_011540381.1 1 - 2 175 107
XM_017002234.1 XP_016857723.1 1 - 2 175 107
GenBank mRNA AK309596.1 - 1 - 2 175 107
BC020702.1 - 1 - 2 174 107
BC132911.1 - 1 - 2 93 107
BC132913.1 - 1 - 2 93 107
M16974.1 - 1 - 2 206 107

View Full Product Details