Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF76
Entrez Gene ID
7629
Gene Name
zinc finger protein 76
Gene Aliases
D6S229E, ZNF523, Zfp523
Chromosome Location
Chr.6: 35259473 - 35295987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292032.1 NP_001278961.1 7 - 8 845 80
NM_003427.4 NP_003418.2 7 - 8 845 80
XM_011514846.2 XP_011513148.1 7 - 8 855 80
XM_011514847.2 XP_011513149.1 7 - 8 993 80
XM_011514849.2 XP_011513151.1 7 - 8 829 80
XM_011514850.2 XP_011513152.1 7 - 8 957 80
XM_011514854.2 XP_011513156.1 7 - 8 842 80
XM_011514855.2 XP_011513157.1 7 - 8 970 80
XM_011514856.2 XP_011513158.1 4 - 5 982 80
XM_017011248.1 XP_016866737.1 7 - 8 993 80
XM_017011249.1 XP_016866738.1 7 - 8 957 80
XM_017011250.1 XP_016866739.1 7 - 8 993 80
XM_017011251.1 XP_016866740.1 7 - 8 993 80
XM_017011252.1 XP_016866741.1 7 - 8 993 80
XM_017011253.1 XP_016866742.1 7 - 8 957 80
XM_017011254.1 XP_016866743.1 7 - 8 829 80
XM_017011255.1 XP_016866744.1 7 - 8 970 80
XM_017011256.1 XP_016866745.1 7 - 8 972 80
XM_017011257.1 XP_016866746.1 7 - 8 970 80
GenBank mRNA AK294605.1 - 7 - 8 829 80
AK295276.1 - 7 - 8 947 80
AK302903.1 - 7 - 8 947 80
BC000133.2 - 7 - 8 825 80
BC002549.2 - 7 - 8 826 80
BT007316.1 - 6 - 7 621 80
M91592.1 - 7 - 8 784 80

View Full Product Details