Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF76
Entrez Gene ID
7629
Gene Name
zinc finger protein 76
Gene Aliases
D6S229E, ZNF523, Zfp523
Chromosome Location
Chr.6: 35259473 - 35295987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292032.1 NP_001278961.1 10 - 11 1389 63
NM_003427.4 NP_003418.2 10 - 11 1389 63
XM_011514846.2 XP_011513148.1 10 - 11 1399 63
XM_011514847.2 XP_011513149.1 10 - 11 1537 63
XM_011514849.2 XP_011513151.1 10 - 11 1373 63
XM_011514850.2 XP_011513152.1 10 - 11 1501 63
XM_011514854.2 XP_011513156.1 10 - 11 1386 63
XM_011514855.2 XP_011513157.1 10 - 11 1514 63
XM_011514856.2 XP_011513158.1 7 - 8 1526 63
XM_017011248.1 XP_016866737.1 10 - 11 1537 63
XM_017011249.1 XP_016866738.1 10 - 11 1501 63
XM_017011250.1 XP_016866739.1 10 - 11 1447 63
XM_017011251.1 XP_016866740.1 10 - 11 1537 63
XM_017011252.1 XP_016866741.1 10 - 11 1537 63
XM_017011253.1 XP_016866742.1 10 - 11 1501 63
XM_017011254.1 XP_016866743.1 10 - 11 1373 63
XM_017011255.1 XP_016866744.1 10 - 11 1514 63
XM_017011256.1 XP_016866745.1 10 - 11 1516 63
XM_017011257.1 XP_016866746.1 10 - 11 1514 63
GenBank mRNA AB208784.1 - 9 - 10 2254 63
BC000133.2 - 10 - 11 1369 63
BC002549.2 - 10 - 11 1370 63
BT007316.1 - 9 - 10 1165 63
M91592.1 - 10 - 11 1328 63

View Full Product Details