The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005698.3 | NP_005689.2 | 7 - 8 | 1015 | 56 |
NM_052837.2 | NP_443069.1 | 6 - 7 | 937 | 56 | |
XM_006711105.3 | XP_006711168.1 | 7 - 8 | 1291 | 56 | |
XM_006711106.3 | XP_006711169.1 | 5 - 6 | 1132 | 56 | |
GenBank mRNA | AF005039.1 | - | 7 - 8 | 877 | 56 |
BC000161.2 | - | 7 - 8 | 889 | 56 | |
BC005135.2 | - | 7 - 8 | 862 | 56 | |
BC010505.1 | - | 6 - 7 | 806 | 56 | |
CR541757.1 | - | 7 - 8 | 781 | 56 |