Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF76
Entrez Gene ID
7629
Gene Name
zinc finger protein 76
Gene Aliases
D6S229E, ZNF523, Zfp523
Chromosome Location
Chr.6: 35259473 - 35295987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292032.1 NP_001278961.1 4 - 5 457 84
NM_003427.4 NP_003418.2 4 - 5 457 84
XM_011514849.2 XP_011513151.1 4 - 5 441 84
XM_011514850.2 XP_011513152.1 4 - 5 569 84
XM_011514854.2 XP_011513156.1 4 - 5 441 84
XM_011514855.2 XP_011513157.1 4 - 5 569 84
XM_017011249.1 XP_016866738.1 4 - 5 569 84
XM_017011253.1 XP_016866742.1 4 - 5 569 84
XM_017011254.1 XP_016866743.1 4 - 5 441 84
XM_017011255.1 XP_016866744.1 4 - 5 569 84
XM_017011256.1 XP_016866745.1 4 - 5 584 84
XM_017011257.1 XP_016866746.1 4 - 5 569 84
GenBank mRNA AB208784.1 - 4 - 5 734 84
AK294605.1 - 4 - 5 441 84
AK295276.1 - 4 - 5 559 84
AK302903.1 - 4 - 5 559 84
AK304626.1 - 4 - 5 808 84
BC000133.2 - 4 - 5 437 84
BC002549.2 - 4 - 5 438 84
BT007316.1 - 3 - 4 233 84
DC299240.1 - 4 - 5 457 84
M91592.1 - 4 - 5 396 84

View Full Product Details