The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017947.2 | NP_060417.2 | 9 - 10 | 1978 | 89 |
GenBank mRNA | AK000740.1 | - | 9 - 10 | 1976 | 89 |
AK222886.1 | - | 9 - 10 | 1965 | 89 | |
AL834481.1 | - | 7 - 8 | 1674 | 89 | |
BC012079.1 | - | 9 - 10 | 1968 | 89 |