The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015117.2 | NP_055932.2 | 9 - 10 | 2342 | 85 |
XM_006716536.3 | XP_006716599.2 | 11 - 12 | 2731 | 85 | |
XM_017013248.1 | XP_016868737.1 | 10 - 11 | 3362 | 85 | |
XM_017013249.1 | XP_016868738.1 | 10 - 11 | 2904 | 85 | |
GenBank mRNA | BC038670.1 | - | 9 - 10 | 2325 | 85 |
D63484.1 | - | 9 - 10 | 2302 | 85 |