Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PRPF4B
- Entrez Gene ID
-
8899
- Gene Name
- pre-mRNA processing factor 4B
- Gene Aliases
-
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1
- Chromosome Location
-
Chr.6: 4019402 - 4064983 on Build GRCh38
View Full Product Details