Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPF4B
Entrez Gene ID
8899
Gene Name
pre-mRNA processing factor 4B
Gene Aliases
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1
Chromosome Location
Chr.6: 4019402 - 4064983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003913.4 NP_003904.3 5 - 6 1738 148
XM_011514970.2 XP_011513272.1 5 - 6 3671 148
XM_017011410.1 XP_016866899.1 5 - 6 3671 148
XM_017011411.1 XP_016866900.1 5 - 6 3671 148
XM_017011412.1 XP_016866901.1 5 - 6 3671 148
XM_017011413.1 XP_016866902.1 5 - 6 3671 148
XM_017011414.1 XP_016866903.1 5 - 6 3671 148
XM_017011415.1 XP_016866904.1 5 - 6 3671 148
XM_017011416.1 XP_016866905.1 5 - 6 3671 148
XM_017011417.1 XP_016866906.1 5 - 6 3671 148
XM_017011418.1 XP_016866907.1 5 - 6 3671 148
XM_017011419.1 XP_016866908.1 5 - 6 3671 148
XM_017011420.1 XP_016866909.1 5 - 6 3671 148
XM_017011421.1 XP_016866910.1 5 - 6 3671 148
XM_017011422.1 XP_016866911.1 5 - 6 3671 148
XM_017011423.1 XP_016866912.1 5 - 6 3671 148
XM_017011424.1 XP_016866913.1 5 - 6 3671 148
XM_017011425.1 XP_016866914.1 5 - 6 3671 148
XM_017011426.1 XP_016866915.1 5 - 6 3671 148
XM_017011427.1 XP_016866916.1 5 - 6 3671 148
XM_017011428.1 XP_016866917.1 5 - 6 3671 148
XM_017011429.1 XP_016866918.1 5 - 6 3671 148
XM_017011430.1 XP_016866919.1 5 - 6 3671 148
XM_017011431.1 XP_016866920.1 5 - 6 3671 148
XM_017011432.1 XP_016866921.1 5 - 6 3671 148
XM_017011433.1 XP_016866922.1 5 - 6 3671 148
XM_017011434.1 XP_016866923.1 5 - 6 3671 148
XM_017011435.1 XP_016866924.1 5 - 6 3671 148
GenBank mRNA AB011108.1 - 5 - 6 1711 148
AF283465.1 - 5 - 6 1689 148
AK291244.1 - 5 - 6 1716 148
AK303047.1 - 5 - 6 1763 148
AY029347.1 - 5 - 6 1647 148
BC009844.2 - 3 - 4 386 148
BC034969.1 - 5 - 6 1751 148
U48736.1 - 2 - 3 227 148

View Full Product Details