Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN4
Entrez Gene ID
23190
Gene Name
UBX domain protein 4
Gene Aliases
UBXD2, UBXDC1, erasin
Chromosome Location
Chr.2: 135741619 - 135785063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014607.3 NP_055422.1 8 - 9 1136 66
GenBank mRNA AK026971.1 - 8 - 9 759 66
AK095402.1 - 4 - 5 524 66
AK292829.1 - 8 - 9 829 66
AK304096.1 - 8 - 9 846 66
BC035594.2 - 9 - 10 968 66
BX647216.1 - 6 - 7 1428 66
BX647662.1 - 8 - 9 928 66
D87684.1 - 8 - 9 889 66
EF199840.1 - 8 - 9 953 66

View Full Product Details