Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf40
Entrez Gene ID
283461
Gene Name
chromosome 12 open reading frame 40
Gene Aliases
HEL-206, HEL-S-94
Chromosome Location
Chr.12: 39626167 - 39908300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031748.3 NP_001026918.2 10 - 11 1514 135
NR_135051.1 10 - 11 1567 135
XM_005268806.3 XP_005268863.1 9 - 10 1324 135
XM_011538231.2 XP_011536533.1 9 - 10 1551 135
GenBank mRNA AK097445.1 - 10 - 11 1516 135
BC048120.1 - 10 - 11 1498 135
EU668334.1 - 10 - 11 1516 135
GQ472216.1 - 10 - 11 1516 135

View Full Product Details