The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031748.3 | NP_001026918.2 | 7 - 8 | 853 | 154 |
NM_001319247.1 | NP_001306176.1 | 7 - 8 | 853 | 154 | |
NR_135051.1 | 6 - 7 | 714 | 154 | ||
XM_005268806.3 | XP_005268863.1 | 6 - 7 | 663 | 154 | |
XM_011538231.2 | XP_011536533.1 | 6 - 7 | 890 | 154 | |
GenBank mRNA | AK097445.1 | - | 6 - 7 | 663 | 154 |
BC038754.1 | - | 7 - 8 | 850 | 154 | |
BC048120.1 | - | 7 - 8 | 837 | 154 | |
EU668334.1 | - | 6 - 7 | 663 | 154 | |
GQ472216.1 | - | 6 - 7 | 663 | 154 |