The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293083.1 | NP_001280012.1 | 11 - 12 | 997 | 78 |
XM_011512110.2 | XP_011510412.1 | 11 - 12 | 1014 | 78 | |
XM_011512111.2 | XP_011510413.1 | 11 - 12 | 1014 | 78 | |
XM_011512113.2 | XP_011510415.1 | 11 - 12 | 1014 | 78 | |
XM_011512114.2 | XP_011510416.1 | 11 - 12 | 1014 | 78 | |
XM_011512115.2 | XP_011510417.1 | 11 - 12 | 1014 | 78 | |
XM_011512116.2 | XP_011510418.1 | 11 - 12 | 1014 | 78 | |
XM_011512117.2 | XP_011510419.1 | 11 - 12 | 1014 | 78 | |
XM_011512119.2 | XP_011510421.1 | 11 - 12 | 1014 | 78 | |
XM_011512120.2 | XP_011510422.1 | 10 - 11 | 905 | 78 | |
XM_011512122.2 | XP_011510424.1 | 11 - 12 | 1015 | 78 | |
XM_011512125.2 | XP_011510427.1 | 11 - 12 | 1015 | 78 | |
XM_011512126.2 | XP_011510428.1 | 11 - 12 | 1014 | 78 | |
XM_011512127.2 | XP_011510429.1 | 11 - 12 | 1015 | 78 | |
XM_011512128.2 | XP_011510430.1 | 11 - 12 | 1014 | 78 | |
XM_017005222.1 | XP_016860711.1 | 11 - 12 | 1014 | 78 | |
GenBank mRNA | DW009988.1 | - | 4 - 5 | 276 | 78 |