Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
Chromosome Location
Chr.2: 96642719 - 96704887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293083.1 NP_001280012.1 28 - 29 3010 115
XM_011512110.2 XP_011510412.1 28 - 29 3027 115
XM_011512111.2 XP_011510413.1 28 - 29 3027 115
XM_011512113.2 XP_011510415.1 28 - 29 3021 115
XM_011512114.2 XP_011510416.1 28 - 29 3027 115
XM_011512115.2 XP_011510417.1 27 - 28 2958 115
XM_011512116.2 XP_011510418.1 28 - 29 2940 115
XM_011512117.2 XP_011510419.1 28 - 29 3027 115
XM_011512118.2 XP_011510420.1 27 - 28 2907 115
XM_011512119.2 XP_011510421.1 28 - 29 3027 115
XM_011512120.2 XP_011510422.1 27 - 28 2918 115
XM_011512122.2 XP_011510424.1 28 - 29 3028 115
XM_011512124.2 XP_011510426.1 12 - 13 2611 115
XM_011512125.2 XP_011510427.1 28 - 29 3028 115
XM_011512126.2 XP_011510428.1 28 - 29 3027 115
XM_011512127.2 XP_011510429.1 28 - 29 3028 115
XM_011512128.2 XP_011510430.1 28 - 29 3027 115
XM_017005222.1 XP_016860711.1 28 - 29 3027 115
XM_017005223.1 XP_016860712.1 11 - 12 1675 115
XM_017005224.1 XP_016860713.1 2 - 3 467 115
GenBank mRNA AL133022.1 - 11 - 12 1369 115
AY461813.1 - 27 - 28 2877 127

View Full Product Details