Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
Chromosome Location
Chr.2: 96642719 - 96704887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293083.1 NP_001280012.1 30 - 31 3310 64
XM_011512110.2 XP_011510412.1 30 - 31 3327 64
XM_011512111.2 XP_011510413.1 30 - 31 3327 64
XM_011512113.2 XP_011510415.1 30 - 31 3321 64
XM_011512114.2 XP_011510416.1 30 - 31 3327 64
XM_011512115.2 XP_011510417.1 29 - 30 3258 64
XM_011512116.2 XP_011510418.1 30 - 31 3240 64
XM_011512117.2 XP_011510419.1 30 - 31 3327 64
XM_011512118.2 XP_011510420.1 29 - 30 3207 64
XM_011512119.2 XP_011510421.1 30 - 31 3327 64
XM_011512120.2 XP_011510422.1 29 - 30 3218 64
XM_011512122.2 XP_011510424.1 30 - 31 3328 64
XM_011512124.2 XP_011510426.1 14 - 15 2911 64
XM_011512125.2 XP_011510427.1 30 - 31 3328 64
XM_011512126.2 XP_011510428.1 30 - 31 3327 64
XM_011512127.2 XP_011510429.1 30 - 31 3328 64
XM_011512128.2 XP_011510430.1 30 - 31 3327 64
XM_017005222.1 XP_016860711.1 30 - 31 3327 64
XM_017005223.1 XP_016860712.1 13 - 14 1975 64
XM_017005224.1 XP_016860713.1 4 - 5 767 64
GenBank mRNA AL133022.1 - 13 - 14 1669 64
AY461813.1 - 29 - 30 3171 64

View Full Product Details