The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293083.1 | NP_001280012.1 | 48 - 49 | 5497 | 85 |
XM_011512110.2 | XP_011510412.1 | 48 - 49 | 5541 | 85 | |
XM_011512111.2 | XP_011510413.1 | 48 - 49 | 5538 | 85 | |
XM_011512113.2 | XP_011510415.1 | 48 - 49 | 5535 | 85 | |
XM_011512114.2 | XP_011510416.1 | 48 - 49 | 5511 | 85 | |
XM_011512115.2 | XP_011510417.1 | 47 - 48 | 5472 | 85 | |
XM_011512116.2 | XP_011510418.1 | 48 - 49 | 5454 | 85 | |
XM_011512117.2 | XP_011510419.1 | 47 - 48 | 5436 | 85 | |
XM_011512118.2 | XP_011510420.1 | 47 - 48 | 5421 | 85 | |
XM_011512119.2 | XP_011510421.1 | 47 - 48 | 5379 | 85 | |
XM_011512120.2 | XP_011510422.1 | 47 - 48 | 5432 | 85 | |
XM_011512122.2 | XP_011510424.1 | 48 - 49 | 5542 | 85 | |
XM_011512124.2 | XP_011510426.1 | 32 - 33 | 5125 | 85 | |
XM_017005222.1 | XP_016860711.1 | 48 - 49 | 5538 | 85 | |
XM_017005223.1 | XP_016860712.1 | 31 - 32 | 4189 | 85 | |
XM_017005224.1 | XP_016860713.1 | 22 - 23 | 2981 | 85 | |
GenBank mRNA | AL133022.1 | - | 30 - 31 | 4900 | 85 |
AY461813.1 | - | 47 - 48 | 5523 | 85 | |
BC117324.1 | - | 14 - 15 | 1705 | 85 | |
BC126368.1 | - | 14 - 15 | 1708 | 85 | |
BC143825.1 | - | 14 - 15 | 1705 | 85 |