Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACSF2
Entrez Gene ID
80221
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
ACSMW, AVYV493
Chromosome Location
Chr.17: 50426158 - 50474845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288968.1 NP_001275897.1 5 - 6 689 103
NM_001288970.1 NP_001275899.1 3 - 4 485 103
NM_001288971.1 NP_001275900.1 2 - 3 289 103
NM_001288972.1 NP_001275901.1 4 - 5 531 103
NM_025149.5 NP_079425.3 4 - 5 614 103
XM_006722110.3 XP_006722173.1 4 - 5 554 103
XM_011525294.1 XP_011523596.1 4 - 5 554 103
GenBank mRNA AK024573.1 - 4 - 5 521 103
AK225044.1 - 4 - 5 521 103
AK225561.1 - 4 - 5 520 103
AK294205.1 - 5 - 6 631 103
AK300145.1 - 3 - 4 393 103
AK300625.1 - 2 - 3 289 103
AY358660.1 - 4 - 5 550 103
BC012053.1 - 4 - 5 527 103
BC014123.2 - 4 - 5 534 103
DC296100.1 - 4 - 5 471 103
DC384989.1 - 2 - 3 289 103

View Full Product Details