Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACSF2
Entrez Gene ID
80221
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
ACSMW, AVYV493
Chromosome Location
Chr.17: 50426158 - 50474845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288968.1 NP_001275897.1 7 - 8 966 112
NM_001288969.1 NP_001275898.1 6 - 7 852 112
NM_001288970.1 NP_001275899.1 5 - 6 762 112
NM_001288971.1 NP_001275900.1 4 - 5 566 112
NM_001288972.1 NP_001275901.1 6 - 7 808 112
NM_025149.5 NP_079425.3 6 - 7 891 112
XM_006722110.3 XP_006722173.1 6 - 7 831 112
XM_011525294.1 XP_011523596.1 6 - 7 831 112
GenBank mRNA AK024573.1 - 6 - 7 798 112
AK225044.1 - 6 - 7 798 112
AK225561.1 - 6 - 7 797 112
AK294205.1 - 7 - 8 908 112
AK295258.1 - 6 - 7 760 112
AK300145.1 - 5 - 6 670 112
AK300625.1 - 4 - 5 566 112
AY358660.1 - 6 - 7 827 112
BC012053.1 - 6 - 7 804 112
BC014123.2 - 6 - 7 811 112

View Full Product Details