The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135820.1 | NP_001129292.1 | 11 - 12 | 2618 | 87 |
NM_013390.2 | NP_037522.1 | 12 - 13 | 2807 | 87 | |
XM_005251869.4 | XP_005251926.1 | 12 - 13 | 2440 | 87 | |
GenBank mRNA | AB037833.2 | - | 12 - 13 | 2400 | 87 |
AF137030.1 | - | 12 - 13 | 2415 | 87 | |
AK075370.1 | - | 5 - 6 | 535 | 87 | |
BC140773.1 | - | 11 - 12 | 2120 | 87 | |
BC144510.1 | - | 12 - 13 | 2275 | 87 | |
BC146780.1 | - | 12 - 13 | 2400 | 87 | |
BC171782.1 | - | 11 - 12 | 2121 | 87 |