Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135820.1 NP_001129292.1 8 - 9 2330 119
NM_013390.2 NP_037522.1 9 - 10 2519 119
XM_005251869.4 XP_005251926.1 9 - 10 2152 119
GenBank mRNA AB037833.2 - 9 - 10 2112 119
AF137030.1 - 9 - 10 2127 119
AK075370.1 - 2 - 3 247 119
BC140773.1 - 8 - 9 1832 119
BC144510.1 - 9 - 10 1987 119
BC146780.1 - 9 - 10 2112 119
BC171782.1 - 8 - 9 1833 119

View Full Product Details