The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018725.3 | NP_061195.2 | 2 - 3 | 131 | 88 |
XM_005265310.4 | XP_005265367.1 | 2 - 3 | 131 | 88 | |
XM_005265311.4 | XP_005265368.1 | 2 - 3 | 156 | 88 | |
XM_005265312.4 | XP_005265369.1 | 2 - 3 | 187 | 88 | |
XM_011533941.2 | XP_011532243.1 | 2 - 3 | 131 | 88 | |
XM_017006804.1 | XP_016862293.1 | 2 - 3 | 128 | 88 | |
XM_017006805.1 | XP_016862294.1 | 2 - 3 | 128 | 88 | |
XM_017006806.1 | XP_016862295.1 | 2 - 3 | 128 | 88 | |
XM_017006807.1 | XP_016862296.1 | 2 - 3 | 131 | 88 | |
GenBank mRNA | AF208110.1 | - | 2 - 3 | 97 | 88 |
AF208111.1 | - | 2 - 3 | 97 | 88 | |
AF212365.1 | - | 2 - 3 | 101 | 88 | |
AF250309.1 | - | 2 - 3 | 92 | 88 | |
AK307749.1 | - | 2 - 3 | 113 | 88 | |
AY359122.1 | - | 2 - 3 | 98 | 88 | |
BC000980.1 | - | 2 - 3 | 101 | 88 | |
DA093617.1 | - | 2 - 3 | 131 | 88 |