The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014227.2 | NP_055042.1 | 8 - 9 | 891 | 77 |
XM_006724308.3 | XP_006724371.1 | 9 - 10 | 1142 | 77 | |
XM_011530342.2 | XP_011528644.1 | 10 - 11 | 1260 | 77 | |
XM_011530343.2 | XP_011528645.1 | 8 - 9 | 806 | 77 | |
XM_011530344.2 | XP_011528646.1 | 7 - 8 | 771 | 77 | |
XM_017028920.1 | XP_016884409.1 | 10 - 11 | 1221 | 77 | |
GenBank mRNA | AJ133127.1 | - | 8 - 9 | 905 | 77 |