The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320606.1 | NP_001307535.1 | 4 - 5 | 1451 | 121 |
NM_001320608.1 | NP_001307537.1 | 3 - 4 | 1471 | 121 | |
NM_002162.4 | NP_002153.2 | 4 - 5 | 1154 | 121 | |
GenBank mRNA | AF340060.1 | - | 4 - 5 | 907 | 121 |
AF340061.1 | - | 4 - 5 | 907 | 121 | |
AK226023.1 | - | 4 - 5 | 988 | 121 | |
AK309643.1 | - | 4 - 5 | 1236 | 121 | |
AK312826.1 | - | 4 - 5 | 989 | 121 | |
AX746321.1 | - | 4 - 5 | 949 | 121 | |
BC040123.1 | - | 4 - 5 | 934 | 121 | |
BC058903.1 | - | 4 - 5 | 940 | 121 | |
S50015.1 | - | 4 - 5 | 935 | 121 | |
X69711.1 | - | 4 - 5 | 949 | 121 | |
X69819.1 | - | 4 - 5 | 942 | 121 |