The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318066.1 | NP_001304995.1 | 13 - 14 | 2012 | 69 |
NM_014822.3 | NP_055637.2 | 13 - 14 | 2009 | 69 | |
XM_005263379.2 | XP_005263436.1 | 13 - 14 | 1886 | 69 | |
XM_017008875.1 | XP_016864364.1 | 8 - 9 | 1167 | 69 | |
GenBank mRNA | AB018298.1 | - | 13 - 14 | 1912 | 69 |
AF130464.2 | - | 12 - 13 | 1712 | 69 | |
AK001390.1 | - | 10 - 11 | 1380 | 69 | |
AK125962.1 | - | 7 - 8 | 1000 | 69 | |
AK291765.1 | - | 13 - 14 | 1909 | 69 | |
AK309960.1 | - | 6 - 7 | 748 | 69 | |
BC035761.1 | - | 13 - 14 | 1934 | 69 | |
BC037736.1 | - | 16 - 17 | 2486 | 69 |