The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318066.1 | NP_001304995.1 | 6 - 7 | 1099 | 66 |
NM_014822.3 | NP_055637.2 | 6 - 7 | 1096 | 66 | |
XM_005263379.2 | XP_005263436.1 | 6 - 7 | 973 | 66 | |
GenBank mRNA | AB018298.1 | - | 6 - 7 | 999 | 66 |
AF130464.2 | - | 5 - 6 | 799 | 66 | |
AK001390.1 | - | 3 - 4 | 467 | 66 | |
AK291765.1 | - | 6 - 7 | 996 | 66 | |
AK302479.1 | - | 7 - 8 | 997 | 66 | |
AK310259.1 | - | 5 - 6 | 810 | 66 | |
BC035761.1 | - | 6 - 7 | 1021 | 66 | |
BC037736.1 | - | 7 - 8 | 1140 | 66 |