The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032287.2 | NP_001027458.1 | 9 - 10 | 1384 | 67 |
NM_001127362.1 | NP_001120834.1 | 9 - 10 | 1384 | 67 | |
NM_003297.3 | NP_003288.2 | 9 - 10 | 1384 | 67 | |
XM_005269133.2 | XP_005269190.1 | 9 - 10 | 1371 | 67 | |
XM_006719584.2 | XP_006719647.1 | 9 - 10 | 1371 | 67 | |
XM_006719585.2 | XP_006719648.1 | 10 - 11 | 1409 | 67 | |
XM_011538716.2 | XP_011537018.1 | 9 - 10 | 1371 | 67 | |
GenBank mRNA | AB307707.1 | - | 8 - 9 | 1128 | 67 |
AK291319.1 | - | 9 - 10 | 1348 | 67 | |
BC040141.1 | - | 9 - 10 | 1336 | 67 | |
JQ308177.1 | - | 8 - 9 | 1128 | 67 | |
JQ308178.1 | - | 8 - 9 | 1128 | 67 | |
M21985.1 | - | 9 - 10 | 1254 | 67 | |
M29959.1 | - | 9 - 10 | 1254 | 67 | |
M29960.1 | - | 9 - 10 | 1184 | 67 |