The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301085.1 | NP_001288014.1 | 2 - 3 | 837 | 107 |
NM_006775.2 | NP_006766.1 | 2 - 3 | 837 | 107 | |
NM_206853.2 | NP_996735.1 | 2 - 3 | 837 | 107 | |
NM_206854.2 | NP_996736.1 | 2 - 3 | 837 | 107 | |
NM_206855.2 | NP_996737.1 | 2 - 3 | 837 | 107 | |
XM_011536258.2 | XP_011534560.1 | 2 - 3 | 344 | 107 | |
XM_011536259.2 | XP_011534561.1 | 2 - 3 | 344 | 107 | |
XM_011536260.2 | XP_011534562.1 | 2 - 3 | 879 | 107 | |
XM_011536261.1 | XP_011534563.1 | 2 - 3 | 344 | 107 | |
XM_017011502.1 | XP_016866991.1 | 2 - 3 | 344 | 107 | |
XM_017011503.1 | XP_016866992.1 | 2 - 3 | 344 | 107 | |
XM_017011504.1 | XP_016866993.1 | 2 - 3 | 344 | 107 | |
GenBank mRNA | AB067798.1 | - | 2 - 3 | 838 | 107 |
AB067799.1 | - | 2 - 3 | 838 | 107 | |
AB067800.1 | - | 2 - 3 | 838 | 107 | |
AB067801.1 | - | 2 - 3 | 838 | 107 | |
AF142422.1 | - | 2 - 3 | 397 | 107 | |
AF272349.1 | - | 2 - 3 | 719 | 107 | |
AK027309.1 | - | 2 - 3 | 244 | 107 | |
AY780788.1 | - | 2 - 3 | 274 | 107 | |
BC012222.1 | - | 1 - 2 | 144 | 107 | |
BC019917.2 | - | 2 - 3 | 325 | 107 | |
DQ323998.1 | - | 2 - 3 | 286 | 107 |