Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5DC3
Entrez Gene ID
51559
Gene Name
5'-nucleotidase domain containing 3
Gene Aliases
TU12B1-TY, TU12B1TY
Chromosome Location
Chr.12: 103746315 - 103841238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031701.2 NP_001026871.1 5 - 6 658 77
XM_011538476.2 XP_011536778.1 2 - 3 415 77
XM_017019454.1 XP_016874943.1 5 - 6 699 77
GenBank mRNA AB032773.1 - 5 - 6 468 77
BC052293.1 - 5 - 6 658 77

View Full Product Details