The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001697.2 | NP_001688.1 | 1 - 2 | 131 | 97 |
GenBank mRNA | AK222608.1 | - | 1 - 2 | 63 | 97 |
AK222962.1 | - | 1 - 2 | 58 | 97 | |
AK311796.1 | - | 1 - 2 | 58 | 97 | |
AV734214.1 | - | 1 - 2 | 131 | 97 | |
BC022865.1 | - | 1 - 2 | 62 | 97 | |
CD174837.1 | - | 1 - 2 | 101 | 97 | |
X83218.1 | - | 1 - 2 | 76 | 97 |