The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030973.3 | NP_112235.2 | 16 - 17 | 2028 | 169 |
XM_011527353.1 | XP_011525655.1 | 16 - 17 | 2020 | 169 | |
GenBank mRNA | AF261072.1 | - | 16 - 17 | 2007 | 169 |
AF283769.2 | - | 11 - 12 | 1488 | 169 | |
AF447873.1 | - | 3 - 4 | 2584 | 169 | |
AK055844.1 | - | 17 - 18 | 1955 | 169 | |
AK289460.1 | - | 16 - 17 | 2018 | 169 | |
AK298897.1 | - | 17 - 18 | 1955 | 169 | |
AY533507.1 | - | 16 - 17 | 1965 | 260 | |
BC021543.1 | - | 15 - 16 | 1808 | 169 | |
BC024312.2 | - | 12 - 13 | 1454 | 169 | |
BC065297.1 | - | 16 - 17 | 2001 | 169 | |
BG770032.1 | - | 1 - 2 | 42 | 169 | |
EU392499.1 | - | 1 - 2 | 79 | 169 | |
EU392500.1 | - | 11 - 12 | 1326 | 169 | |
EU392501.1 | - | 10 - 11 | 1170 | 169 | |
EU392508.1 | - | 3 - 4 | 361 | 169 | |
EU392510.1 | - | 3 - 4 | 264 | 169 | |
EU392511.1 | - | 4 - 5 | 255 | 169 | |
EU392512.1 | - | 3 - 4 | 228 | 169 | |
EU392513.1 | - | 2 - 3 | 165 | 169 | |
EU392514.1 | - | 2 - 3 | 117 | 169 | |
EU392519.1 | - | 9 - 10 | 904 | 169 |