The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201359.1 | NP_001188288.1 | 2 - 3 | 187 | 101 |
NM_005624.3 | NP_005615.2 | 2 - 3 | 187 | 101 | |
XM_011528177.2 | XP_011526479.1 | 3 - 4 | 388 | 101 | |
XM_017027119.1 | XP_016882608.1 | 3 - 4 | 354 | 101 | |
XM_017027120.1 | XP_016882609.1 | 3 - 4 | 283 | 101 | |
XM_017027121.1 | XP_016882610.1 | 3 - 4 | 229 | 101 | |
XM_017027122.1 | XP_016882611.1 | 3 - 4 | 366 | 101 | |
GenBank mRNA | AB046579.1 | - | 1 - 2 | 76 | 101 |
AK292662.1 | - | 2 - 3 | 187 | 101 | |
BC125136.1 | - | 1 - 2 | 80 | 101 | |
BC130561.1 | - | 1 - 2 | 122 | 101 | |
BC144463.1 | - | 1 - 2 | 122 | 101 | |
U86358.1 | - | 1 - 2 | 76 | 101 |