Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKG2
Entrez Gene ID
5593
Gene Name
protein kinase, cGMP-dependent, type II
Gene Aliases
PRKGR2, cGK2, cGKII
Chromosome Location
Chr.4: 81086874 - 81217837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282485.1 NP_001269414.1 2 - 3 652 135
NM_006259.2 NP_006250.1 2 - 3 652 135
XM_005263126.3 XP_005263183.1 2 - 3 1087 135
XM_017008413.1 XP_016863902.1 2 - 3 1089 135
XM_017008414.1 XP_016863903.1 2 - 3 1084 135
XM_017008415.1 XP_016863904.1 2 - 3 494 135
XM_017008416.1 XP_016863905.1 2 - 3 1006 135
XM_017008417.1 XP_016863906.1 1 - 2 470 135
XM_017008418.1 XP_016863907.1 1 - 2 470 135
GenBank mRNA AK297673.1 - 2 - 3 599 135
D70899.1 - 1 - 2 457 135
X94612.1 - 1 - 2 471 135

View Full Product Details