Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADH4
Entrez Gene ID
127
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Gene Aliases
ADH-2, HEL-S-4
Chromosome Location
Chr.4: 99123657 - 99144330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000670.4 NP_000661.2 2 - 3 195 100
NM_001306171.1 NP_001293100.1 3 - 4 343 100
NM_001306172.1 NP_001293101.1 3 - 4 335 100
XM_017007713.1 XP_016863202.1 3 - 4 366 100
XM_017007714.1 XP_016863203.1 3 - 4 374 100
XM_017007715.1 XP_016863204.1 2 - 3 227 100
GenBank mRNA AK290835.1 - 2 - 3 194 100
AK295556.1 - 3 - 4 342 100
AK313649.1 - 2 - 3 194 100
AV659353.1 - 2 - 3 195 100
AV659364.1 - 2 - 3 195 100
BC022319.1 - 2 - 3 194 100
CD014080.1 - 3 - 4 313 100

View Full Product Details