The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190233.1 | NP_001177162.1 | 2 - 3 | 239 | 77 |
NM_145261.3 | NP_660304.1 | 2 - 3 | 227 | 77 | |
NR_033722.1 | 2 - 3 | 227 | 77 | ||
NR_033723.1 | 2 - 3 | 227 | 77 | ||
GenBank mRNA | AI268262.1 | - | 2 - 3 | 226 | 77 |
AK307097.1 | - | 2 - 3 | 216 | 77 | |
AK311765.1 | - | 2 - 3 | 155 | 77 | |
AW161751.1 | - | 4 - 5 | 77 | ||
BC009702.1 | - | 2 - 3 | 179 | 77 | |
BC073989.1 | - | 2 - 3 | 172 | 77 | |
BE773017.1 | - | 2 - 3 | 194 | 77 | |
BF666599.1 | - | 2 - 3 | 189 | 77 | |
BM701756.1 | - | 2 - 3 | 180 | 77 | |
BM702203.1 | - | 2 - 3 | 180 | 77 | |
CB052107.1 | - | 2 - 3 | 165 | 77 | |
CB159371.1 | - | 2 - 3 | 239 | 77 | |
KU178861.1 | - | 1 - 2 | 55 | 77 | |
KU178862.1 | - | 1 - 2 | 55 | 77 |