Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHISA6
Entrez Gene ID
388336
Gene Name
shisa family member 6
Gene Aliases
-
Chromosome Location
Chr.17: 11241423 - 11564063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL110201.1 - 1 - 1 1263 117


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream phosphoinositide interacting regulator of transient receptor potential channels PIRT -450.1kb Anti-sense Chr. - 10822475 - 10838101 on Build GRCh38
Downstream dynein axonemal heavy chain 9 DNAH9 DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20 307.6kb Sense Chr. - 11598431 - 11970168 on Build GRCh38

View Full Product Details