Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENAM
Entrez Gene ID
10117
Gene Name
enamelin
Gene Aliases
ADAI, AI1C, AIH2
Chromosome Location
Chr.4: 70627471 - 70646819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031889.2 NP_114095.2 1 - 2 220 200
GenBank mRNA AF125373.1 - 1 - 2 220 200

View Full Product Details