Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFAP46
Entrez Gene ID
54777
Gene Name
cilia and flagella associated protein 46
Gene Aliases
C10orf123, C10orf124, C10orf92, C10orf93, TTC40, bA288G11.4, bA288G11.5, bB137A17.2, bB137A17.3
Chromosome Location
Chr.10: 132808392 - 132942585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001200049.2 NP_001186978.2 53 - 54 7385 65
GenBank mRNA AL122111.1 - 10 - 11 886 65
BC034223.1 - 15 - 16 1853 65

View Full Product Details