Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 13 - 14 1687 111
NM_020784.2 NP_065835.2 13 - 14 1702 111
GenBank mRNA AB037765.2 - 13 - 14 1614 111
BC137081.1 - 13 - 14 1582 111
BC137082.1 - 13 - 14 1582 111
BC142650.1 - 13 - 14 1625 111
BC144469.1 - 13 - 14 1567 111
BC150205.1 - 13 - 14 1623 111
BC152428.1 - 13 - 14 1614 111

View Full Product Details