Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 1 - 2 270 169
NM_020784.2 NP_065835.2 1 - 2 270 169
XM_017021505.1 XP_016876994.1 1 - 2 250 169
GenBank mRNA AB037765.2 - 1 - 2 182 169
BC137081.1 - 1 - 2 150 169
BC137082.1 - 1 - 2 150 169
BC142650.1 - 1 - 2 193 169
BC144469.1 - 1 - 2 150 169
BC150205.1 - 1 - 2 191 169
BC152428.1 - 1 - 2 182 169
DB083799.1 - 1 - 2 250 169

View Full Product Details