Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 20 - 21 2633 130
NM_020784.2 NP_065835.2 20 - 21 2648 130
GenBank mRNA AB037765.2 - 20 - 21 2560 130
AK022563.1 - 6 - 7 834 130
AK097748.1 - 21 - 22 2537 130
BC137081.1 - 20 - 21 2528 130
BC137082.1 - 20 - 21 2528 130
BC142650.1 - 20 - 21 2571 130
BC144469.1 - 20 - 21 2513 130
BC150205.1 - 20 - 21 2569 130
BC152428.1 - 20 - 21 2560 130

View Full Product Details