The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016337.2 | NP_057421.1 | 4 - 5 | 511 | 96 |
XM_005267749.3 | XP_005267806.1 | 4 - 5 | 1130 | 96 | |
XM_011536828.2 | XP_011535130.1 | 4 - 5 | 1144 | 96 | |
XM_017021363.1 | XP_016876852.1 | 4 - 5 | 1130 | 96 | |
XM_017021364.1 | XP_016876853.1 | 4 - 5 | 1017 | 96 | |
GenBank mRNA | AF052504.1 | - | 4 - 5 | 485 | 96 |
AF087843.1 | - | 4 - 5 | 490 | 96 | |
AF112209.1 | - | 4 - 5 | 604 | 96 | |
AF131766.1 | - | 2 - 3 | 258 | 96 | |
AK289720.1 | - | 5 - 6 | 521 | 96 | |
AK295919.1 | - | 4 - 5 | 719 | 96 | |
AK301379.1 | - | 4 - 5 | 503 | 96 | |
BC023997.2 | - | 4 - 5 | 660 | 96 | |
BC032358.1 | - | 4 - 5 | 482 | 96 |