Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAGE3
Entrez Gene ID
139793
Gene Name
PAGE family member 3
Gene Aliases
CT16.6, GAGED1, PAGE-3
Chromosome Location
Chr.X: 55257833 - 55264916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017931.2 NP_001017931.2 4 - 5 629 86
NM_001171252.1 NP_001164723.1 4 - 5 643 86
NM_001303613.1 NP_001290542.1 4 - 5 431 86
XM_017029282.1 XP_016884771.1 4 - 5 432 86
GenBank mRNA BC140001.1 - 4 - 5 502 86
BC140741.1 - 4 - 5 515 86
BC144449.1 - 4 - 5 528 86

View Full Product Details