The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270419.1 | NP_001257348.1 | 3 - 4 | 327 | 98 |
NM_181581.2 | NP_853559.1 | 3 - 4 | 477 | 98 | |
NR_073002.1 | 3 - 4 | 515 | 98 | ||
NR_073003.1 | 3 - 4 | 477 | 98 | ||
NR_073004.1 | 3 - 4 | 477 | 98 | ||
NR_073005.1 | 3 - 4 | 327 | 98 | ||
XM_005250125.4 | XP_005250182.1 | 2 - 3 | 339 | 98 | |
XM_011515766.2 | XP_011514068.1 | 3 - 4 | 471 | 98 | |
XM_011515767.2 | XP_011514069.1 | 2 - 3 | 221 | 98 | |
XM_017011708.1 | XP_016867197.1 | 3 - 4 | 492 | 98 | |
GenBank mRNA | AK294095.1 | - | 3 - 4 | 476 | 98 |
AK297195.1 | - | 3 - 4 | 484 | 98 | |
AL523141.3 | - | 3 - 4 | 447 | 98 | |
BC111774.1 | - | 2 - 3 | 813 | 98 | |
BG250736.1 | - | - | 303 | 98 | |
BX395100.2 | - | 3 - 4 | 286 | 98 | |
CX787147.1 | - | 1 - 2 | 46 | 98 | |
DA223301.1 | - | 3 - 4 | 299 | 98 | |
U62767.1 | - | 3 - 4 | 275 | 98 |