The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_073003.1 | 4 - 5 | 600 | 63 | |
NR_073004.1 | 4 - 5 | 600 | 63 | ||
NR_073005.1 | 4 - 5 | 450 | 63 | ||
GenBank mRNA | AK127802.1 | - | 1 - 2 | 3642 | 63 |
AK294095.1 | - | 4 - 5 | 599 | 63 | |
AL523141.3 | - | 4 - 5 | 570 | 63 | |
BC111774.1 | - | 3 - 4 | 936 | 63 | |
DA223301.1 | - | 4 - 5 | 422 | 63 |